siRNA Details
| Gene Symbol: Faim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144756.2 | NP_653357.1 | 12 | 966 | ||
| GenBank mRNA | AF044201.1 | - | 12 | 915 | ||
| BC087606.1 | - | 12 | 966 | 7314068 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Faim2| Gene Symbol: Faim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144756.2 | NP_653357.1 | 12 | 966 | ||
| GenBank mRNA | AF044201.1 | - | 12 | 915 | ||
| BC087606.1 | - | 12 | 966 | 7314068 | ||