siRNA Details
| Gene Symbol: Optn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145081.3 | NP_659549.2 | 2 | 208 | ||
| XM_006254262.2 | XP_006254324.1 | 3 | 366 | |||
| XM_006254263.2 | XP_006254325.1 | 3 | 365 | |||
| XM_006254264.2 | XP_006254326.1 | 3 | 365 | |||
| XM_008771852.1 | XP_008770074.1 | 3 | 366 | |||
| XM_008771853.1 | XP_008770075.1 | 3 | 365 | |||
| GenBank mRNA | AB069907.3 | - | 2 | 208 | ||
| AB222074.1 | - | 2 | 162 | |||
| BC086976.1 | - | 3 | 294 | 7111026 | ||
| FQ223875.1 | - | 3 | 365 | |||

Optn
Genomic Map