siRNA Details
| Gene Symbol: Lrrc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145083.1 | NP_659551.1 | 1 | 754 | ||
| XM_006248482.2 | XP_006248544.1 | 2 | 870 | |||
| GenBank mRNA | AB071036.1 | - | 1 | 754 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc15| Gene Symbol: Lrrc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145083.1 | NP_659551.1 | 1 | 754 | ||
| XM_006248482.2 | XP_006248544.1 | 2 | 870 | |||
| GenBank mRNA | AB071036.1 | - | 1 | 754 | ||