siRNA Details
| Gene Symbol: Myocd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182667.2 | NP_872608.1 | 7 | 860 | ||
| XM_006246582.2 | XP_006246644.1 | 7 | 3301 | |||
| XM_006246583.2 | XP_006246645.1 | 8 | 3345 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myocd| Gene Symbol: Myocd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182667.2 | NP_872608.1 | 7 | 860 | ||
| XM_006246582.2 | XP_006246644.1 | 7 | 3301 | |||
| XM_006246583.2 | XP_006246645.1 | 8 | 3345 | |||