siRNA Details
Gene Symbol: Retsat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145084.1 | NP_659552.1 | 4 | 773 | ||
GenBank mRNA | AF465614.1 | - | 4 | 773 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Retsat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145084.1 | NP_659552.1 | 4 | 773 | ||
GenBank mRNA | AF465614.1 | - | 4 | 773 |