siRNA Details
| Gene Symbol: Serbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145086.1 | NP_659554.1 | 3 | 643 | ||
| XM_006236616.1 | XP_006236678.1 | 3 | 691 | |||
| XM_006236617.1 | XP_006236679.1 | 3 | 691 | |||
| XM_006236618.1 | XP_006236680.1 | 3 | 691 | |||
| GenBank mRNA | AF388527.1 | - | 3 | 649 | ||
| BC079337.1 | - | 3 | 643 | 7106027 | ||
| FQ213573.1 | - | 3 | 686 | |||
| FQ223736.1 | - | 3 | 664 | |||
| FQ225988.1 | - | 3 | 358 | |||
| FQ226374.1 | - | 3 | 664 | |||
| FQ226420.1 | - | 3 | 358 | |||
| FQ227636.1 | - | 3 | 665 | |||
| FQ228026.1 | - | 3 | 654 | |||
| FQ230135.1 | - | 3 | 670 | |||
| FQ230192.1 | - | 3 | 654 | |||
| FQ230382.1 | - | 3 | 357 | |||
| FQ231329.1 | - | 3 | 358 | |||
| FQ231840.1 | - | 3 | 358 | |||
| FQ232288.1 | - | 3 | 664 | |||
| FQ232894.1 | - | 3 | 665 | |||
| FQ233179.1 | - | 3 | 664 | |||
| FQ233457.1 | - | 3 | 670 | |||
| FQ233911.1 | - | 3 | 664 | |||
| FQ234284.1 | - | 3 | 638 | |||
| FQ234534.1 | - | 3 | 665 | |||
| FQ235054.1 | - | 3 | 358 | |||
| U21718.1 | - | 2 | 241 | |||

Serbp1
Genomic Map