siRNA Details
| Gene Symbol: Aard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145093.1 | NP_659561.1 | 2 | 398 | ||
| GenBank mRNA | AY007690.1 | - | 2 | 398 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aard| Gene Symbol: Aard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145093.1 | NP_659561.1 | 2 | 398 | ||
| GenBank mRNA | AY007690.1 | - | 2 | 398 | ||