siRNA Details
| Gene Symbol: Zdhhc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145096.2 | NP_659564.2 | 3 | 460 | ||
| XM_008771288.1 | XP_008769510.1 | 3 | 490 | |||
| GenBank mRNA | AB217870.1 | - | 3 | 259 | ||
| AF228917.1 | - | 3 | 449 | |||
| AY886521.1 | - | 3 | 460 | |||

Zdhhc2
Genomic Map