siRNA Details
| Gene Symbol: Prim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ011608.1 | - | 9, 10 | 985 | ||
| BC088272.1 | - | 9, 10 | 992 | 7379940 | ||
| FQ231746.1 | - | 2, 3 | 191 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prim1| Gene Symbol: Prim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ011608.1 | - | 9, 10 | 985 | ||
| BC088272.1 | - | 9, 10 | 992 | 7379940 | ||
| FQ231746.1 | - | 2, 3 | 191 | |||