siRNA Details
| Gene Symbol: Bcas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145670.4 | NP_663703.3 | 7 | 1157 | ||
| XM_008762466.1 | XP_008760688.1 | 8 | 1292 | |||
| GenBank mRNA | AB029897.1 | - | 6 | 800 | ||
| AB180269.1 | - | 6 | 800 | |||
| AB180270.1 | - | 6 | 800 | |||
| AB180272.1 | - | 5 | 665 | |||
| BC094306.1 | - | 6 | 788 | |||
| BC103480.1 | - | 6 | 1055 | |||
| CO405877.1 | - | 4 | 386 | |||

Bcas1
Genomic Map