siRNA Details
| Gene Symbol: Mafk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145673.2 | NP_663706.2 | 3 | 1579 | ||
| XM_006248915.2 | XP_006248977.1 | 3 | 4820 | |||
| XM_006248917.2 | XP_006248979.1 | 3 | 1453 | |||
| GenBank mRNA | BC070906.1 | - | 3 | 1579 | 7103075 | |

Mafk
Genomic Map