siRNA Details
| Gene Symbol: Mafk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145673.2 | NP_663706.2 | 3 | 613 | ||
| XM_006248915.2 | XP_006248977.1 | 3 | 3854 | |||
| XM_006248917.2 | XP_006248979.1 | 3 | 487 | |||
| GenBank mRNA | AF461686.1 | - | 3 | 528 | ||
| BC070906.1 | - | 3 | 613 | 7103075 | ||

Mafk
Genomic Map