siRNA Details
| Gene Symbol: Gdf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170350.1 | NP_001163821.1 | 2 | 1576 | ||
| XM_006239878.2 | XP_006239940.1 | 2 | 1681 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gdf7| Gene Symbol: Gdf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170350.1 | NP_001163821.1 | 2 | 1576 | ||
| XM_006239878.2 | XP_006239940.1 | 2 | 1681 | |||