siRNA Details
| Gene Symbol: Tcf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001032397.1 | NP_001027569.1 | 2 | 713 | ||
| GenBank mRNA | BC100106.1 | - | 2 | 713 | 7379660 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcf21| Gene Symbol: Tcf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001032397.1 | NP_001027569.1 | 2 | 713 | ||
| GenBank mRNA | BC100106.1 | - | 2 | 713 | 7379660 | |