siRNA Details
Gene Symbol: Rapgef4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761973.1 | XP_008760195.1 | 11 | 1088 | ||
GenBank mRNA | AB031229.1 | - | 11 | 1264 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rapgef4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761973.1 | XP_008760195.1 | 11 | 1088 | ||
GenBank mRNA | AB031229.1 | - | 11 | 1264 |