siRNA Details
| Gene Symbol: Frmd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749805.3 | XP_003749853.1 | 10 | 1862 | ||
| XM_006236974.2 | XP_006237036.1 | 10 | 2534 | |||
| XM_006236975.2 | XP_006237037.1 | 6 | 1484 | |||
| XM_232212.7 | XP_232212.7 | 10 | 843 | |||
| GenBank mRNA | AF087945.1 | - | 5 | 232 | ||

Frmd4b
Genomic Map