siRNA Details
| Gene Symbol: Frmd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749805.3 | XP_003749853.1 | 22 | 4007 | ||
| XM_006236974.2 | XP_006237036.1 | 22 | 4679 | |||
| XM_006236975.2 | XP_006237037.1 | 18 | 3629 | |||
| XM_232212.7 | XP_232212.7 | 22 | 2988 | |||

Frmd4b
Genomic Map