siRNA Details
| Gene Symbol: Zfp394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145724.1 | NP_663776.1 | 3 | 1682 | ||
| GenBank mRNA | AF052042.1 | - | 3 | 1682 | ||
| BC100140.1 | - | 3 | 1676 | 7442414 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp394| Gene Symbol: Zfp394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145724.1 | NP_663776.1 | 3 | 1682 | ||
| GenBank mRNA | AF052042.1 | - | 3 | 1682 | ||
| BC100140.1 | - | 3 | 1676 | 7442414 | ||