siRNA Details
Gene Symbol: Tnfsf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145765.1 | NP_665708.1 | 4 | 319 | ||
GenBank mRNA | AF520787.1 | - | 4 | 319 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tnfsf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145765.1 | NP_665708.1 | 4 | 319 | ||
GenBank mRNA | AF520787.1 | - | 4 | 319 |