siRNA Details
| Gene Symbol: THPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000460.3 | NP_000451.1 | 6 | 859 | ||
| NM_001177597.2 | NP_001171068.1 | 6 | 847 | |||
| NM_001177598.2 | NP_001171069.1 | 6 | 842 | |||
| NM_001289998.1 | NP_001276927.1 | 7 | 1126 | |||
| NM_001290003.1 | NP_001276932.1 | 7 | 1126 | |||
| NM_001290022.1 | NP_001276951.1 | 7 | 1114 | |||
| NM_001290026.1 | NP_001276955.1 | 7 | 1109 | |||
| NM_001290028.1 | NP_001276957.1 | 6 | 823 | |||
| GenBank mRNA | AB102890.1 | - | 4 | 563 | ||
| BC130322.1 | - | 6 | 779 | 40146353 | ||
| BC136338.1 | - | 6 | 791 | 9020325 | ||
| BC143982.1 | - | 6 | 779 | 9052500 | ||
| D32047.1 | - | 5 | 681 | |||
| L33410.1 | - | 6 | 795 | |||
| L36052.1 | - | 4 | 580 | |||
| U11025.1 | - | 5 | 615 | |||
| U59493.1 | - | 4 | 580 | |||
| U59494.1 | - | 4 | 580 | |||

THPO
Genomic Map