siRNA Details
| Gene Symbol: Sra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183329.3 | NP_899158.3 | 5 | 607 | ||
| GenBank mRNA | AY026354.1 | - | 5 | 555 | ||
| AY542868.1 | - | 5 | 604 | |||
| BC091105.1 | - | 4 | 1391 | 7381969 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sra1| Gene Symbol: Sra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183329.3 | NP_899158.3 | 5 | 607 | ||
| GenBank mRNA | AY026354.1 | - | 5 | 555 | ||
| AY542868.1 | - | 5 | 604 | |||
| BC091105.1 | - | 4 | 1391 | 7381969 | ||