siRNA Details
| Gene Symbol: Sra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183329.3 | NP_899158.3 | 4 | 474 | ||
| GenBank mRNA | AY026354.1 | - | 4 | 422 | ||
| AY542868.1 | - | 4 | 471 | |||
| BC091105.1 | - | 3 | 1258 | 7381969 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sra1| Gene Symbol: Sra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183329.3 | NP_899158.3 | 4 | 474 | ||
| GenBank mRNA | AY026354.1 | - | 4 | 422 | ||
| AY542868.1 | - | 4 | 471 | |||
| BC091105.1 | - | 3 | 1258 | 7381969 | ||