siRNA Details
| Gene Symbol: Miox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145771.2 | NP_665714.2 | 7, 8 | 597 | ||
| XM_008765674.1 | XP_008763896.1 | 7, 8 | 781 | |||
| GenBank mRNA | AF197128.1 | - | 7, 8 | 591 | ||
| AF230096.1 | - | 6, 7 | 599 | |||
| AY738259.1 | - | 6, 7 | 591 | |||
| BC078840.1 | - | 7, 8 | 597 | 7110224 | ||
| FQ219187.1 | - | 6, 7 | 586 | |||

Miox
Genomic Map