siRNA Details
| Gene Symbol: Miox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145771.2 | NP_665714.2 | 9, 10 | 766 | ||
| XM_008765674.1 | XP_008763896.1 | 9, 10 | 950 | |||
| GenBank mRNA | BC078840.1 | - | 9, 10 | 766 | 7110224 | |
| FQ219187.1 | - | 8, 9 | 755 | |||

Miox
Genomic Map