siRNA Details
| Gene Symbol: Dgat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012345.1 | NP_001012345.1 | 7 | 1004 | ||
| XM_006229733.2 | XP_006229795.1 | 6 | 1044 | |||
| XM_008759689.1 | XP_008757911.1 | 7 | 755 | |||
| GenBank mRNA | AJ487787.1 | - | 3 | 300 | ||
| BC089846.1 | - | 7 | 1004 | 7372077 | ||

Dgat2
Genomic Map