siRNA Details
| Gene Symbol: Nr1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113422.1 | NP_001106893.1 | 7 | 1698 | ||
| NM_145775.2 | NP_665718.2 | 7 | 1668 | |||
| GenBank mRNA | BC062047.1 | - | 7 | 1698 | 5598020 | |
| DN948099.1 | - | 3 | 772 | |||
| M25804.1 | - | 7 | 1668 | |||

Nr1d1
Genomic Map