siRNA Details
| Gene Symbol: Spag5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044224.1 | NP_001037689.1 | 23 | 3539 | ||
| GenBank mRNA | AB231661.1 | - | 23 | 3539 | ||
| AF111111.1 | - | 11 | 1111 | |||
| BC166408.1 | - | 23 | 3529 | 9030944 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spag5| Gene Symbol: Spag5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044224.1 | NP_001037689.1 | 23 | 3539 | ||
| GenBank mRNA | AB231661.1 | - | 23 | 3539 | ||
| AF111111.1 | - | 11 | 1111 | |||
| BC166408.1 | - | 23 | 3529 | 9030944 | ||