siRNA Details
| Gene Symbol: Olfm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145777.1 | NP_665720.1 | 5 | 827 | ||
| GenBank mRNA | AF442822.1 | - | 5 | 827 | ||
| AF442823.1 | - | 718 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfm3| Gene Symbol: Olfm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145777.1 | NP_665720.1 | 5 | 827 | ||
| GenBank mRNA | AF442822.1 | - | 5 | 827 | ||
| AF442823.1 | - | 718 | ||||