siRNA Details
Gene Symbol: Nr2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145780.1 | NP_665723.1 | 13 | 1738 | ||
GenBank mRNA | BC061822.1 | - | 14 | 1819 | 5599041 | |
L26398.1 | - | 13 | 1738 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nr2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145780.1 | NP_665723.1 | 13 | 1738 | ||
GenBank mRNA | BC061822.1 | - | 14 | 1819 | 5599041 | |
L26398.1 | - | 13 | 1738 |