siRNA Details
| Gene Symbol: Timm13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145781.2 | NP_665724.1 | 3 | 691 | ||
| GenBank mRNA | AF144701.1 | - | 3 | 691 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Timm13| Gene Symbol: Timm13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145781.2 | NP_665724.1 | 3 | 691 | ||
| GenBank mRNA | AF144701.1 | - | 3 | 691 | ||