siRNA Details
| Gene Symbol: Cox5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145783.1 | NP_665726.1 | 4 | 393 | ||
| GenBank mRNA | FQ210272.1 | - | 4 | 426 | ||
| FQ211124.1 | - | 4 | 453 | |||
| FQ217304.1 | - | 4 | 426 | |||
| FQ217698.1 | - | 4 | 416 | |||
| FQ223560.1 | - | 4 | 426 | |||
| FQ223883.1 | - | 4 | 410 | |||
| FQ224560.1 | - | 4 | 418 | |||
| FQ224768.1 | - | 4 | 419 | |||
| FQ228531.1 | - | 4 | 426 | |||
| X15030.1 | - | 4 | 393 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_007747, BC034302, X15963, AK018723.

Cox5a
Genomic Map