siRNA Details
| Gene Symbol: Gpr37l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145784.2 | NP_665727.2 | 1 | 131 | ||
| GenBank mRNA | AF087947.1 | - | 197 | |||
| BC168215.1 | - | 1 | 131 | 7313732 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr37l1| Gene Symbol: Gpr37l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145784.2 | NP_665727.2 | 1 | 131 | ||
| GenBank mRNA | AF087947.1 | - | 197 | |||
| BC168215.1 | - | 1 | 131 | 7313732 | ||
Set Membership: |
|