siRNA Details
| Gene Symbol: Ppp1r32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145786.1 | NP_665729.1 | 6 | 655 | ||
| XM_006231016.2 | XP_006231078.1 | 5 | 547 | |||
| XM_008760216.1 | XP_008758438.1 | 6 | 761 | |||
| GenBank mRNA | AY032664.1 | - | 5 | 519 | ||
| AY032665.1 | - | 6 | 655 | |||
| BC081718.1 | - | 6 | 646 | 7112534 | ||

Ppp1r32
Genomic Map