siRNA Details
| Gene Symbol: Dnah12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252671.2 | XP_006252733.1 | 26 | 3924 | ||
| XM_006252672.2 | XP_006252734.1 | 25 | 3798 | |||
| XM_006252673.2 | XP_006252735.1 | 25 | 3765 | |||
| XM_006252674.2 | XP_006252736.1 | 26 | 3924 | |||
| XM_008771017.1 | XP_008769239.1 | 26 | 5727 | |||
| XM_008771018.1 | XP_008769240.1 | 16 | 2539 | |||
| XM_008771019.1 | XP_008769241.1 | 26 | 3924 | |||
| XM_008771020.1 | XP_008769242.1 | 26 | 3924 | |||

Dnah12
Genomic Map