siRNA Details
| Gene Symbol: Tank | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164073.1 | NP_001157545.1 | 7 | 672 | ||
| NM_145788.2 | NP_665731.2 | 7 | 683 | |||
| XM_006234273.2 | XP_006234335.1 | 7 | 745 | |||
| XM_006234274.2 | XP_006234336.1 | 7 | 731 | |||
| GenBank mRNA | AJ422211.1 | - | 6 | 543 | ||
| BC088854.1 | - | 8 | 751 | |||
| BC129086.1 | - | 7 | 672 | 7365827 | ||
| EV776506.1 | - | 7 | 682 | |||

Tank
Genomic Map