siRNA Details
| Gene Symbol: THRSP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003251.3 | NP_003242.1 | 1 | 290 | ||
| GenBank mRNA | AK311975.1 | - | 1 | 290 | ||
| BC031989.1 | - | 1 | 273 | 4745757 | ||
| BG674197.1 | - | 1 | 243 | |||
| BP317892.1 | - | 1 | 290 | |||
| CB122257.1 | - | 1 | 290 | |||

THRSP
Genomic Map