siRNA Details
| Gene Symbol: Lipogenin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145790.1 | NP_665733.1 | 1 | 1027 | ||
| GenBank mRNA | AY057076.1 | - | 1 | 1027 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lipogenin| Gene Symbol: Lipogenin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145790.1 | NP_665733.1 | 1 | 1027 | ||
| GenBank mRNA | AY057076.1 | - | 1 | 1027 | ||