siRNA Details
| Gene Symbol: Lhx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145880.3 | NP_665887.3 | 3 | 1204 | ||
| GenBank mRNA | FQ213041.1 | - | 2 | 474 | ||
| S71523.1 | - | 3 | 1204 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lhx1| Gene Symbol: Lhx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145880.3 | NP_665887.3 | 3 | 1204 | ||
| GenBank mRNA | FQ213041.1 | - | 2 | 474 | ||
| S71523.1 | - | 3 | 1204 | |||