siRNA Details
| Gene Symbol: LOC257643 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147137.3 | NP_671478.2 | 2 | 290 | ||
| GenBank mRNA | AF442205.1 | - | 2 | 320 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC257643| Gene Symbol: LOC257643 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147137.3 | NP_671478.2 | 2 | 290 | ||
| GenBank mRNA | AF442205.1 | - | 2 | 320 | ||