siRNA Details
| Gene Symbol: Zwint | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147138.2 | NP_671479.1 | 3 | 267 | ||
| GenBank mRNA | AF439397.1 | - | 3 | 249 | ||
| AF532776.1 | - | 3 | 267 | |||
| BC063144.1 | - | 3 | 259 | 6921645 | ||
| EU000468.1 | - | 3 | 235 | |||

Zwint
Genomic Map