siRNA Details
| Gene Symbol: Cacna1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195199.1 | NP_001182128.1 | 32, 34 | 4720 | ||
| NM_147141.1 | NP_671482.1 | 31, 33 | 4748 | |||
| XM_006233569.2 | XP_006233631.1 | 33, 35 | 4866 | |||
| XM_006233570.2 | XP_006233632.1 | 33, 35 | 4863 | |||
| XM_006233573.2 | XP_006233635.1 | 32, 34 | 4854 | |||
| XM_006233575.2 | XP_006233637.1 | 32, 34 | 4803 | |||
| XM_006233576.2 | XP_006233638.1 | 32, 34 | 4800 | |||
| XM_008761578.1 | XP_008759800.1 | 33, 35 | 4866 | |||
| GenBank mRNA | AF055477.2 | - | 31, 33 | 4748 | ||
| HQ008360.1 | - | 32, 34 | 4720 | |||

Cacna1b
Genomic Map