siRNA Details
| Gene Symbol: Cenpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100827.1 | NP_001094297.1 | 12 | 2444 | ||
| GenBank mRNA | AF370442.1 | - | 1 | 199 | ||
| FQ225482.1 | - | 1 | 269 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cenpf| Gene Symbol: Cenpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100827.1 | NP_001094297.1 | 12 | 2444 | ||
| GenBank mRNA | AF370442.1 | - | 1 | 199 | ||
| FQ225482.1 | - | 1 | 269 | |||