siRNA Details
Gene Symbol: Dclre1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147145.2 | NP_671486.2 | 8 | 658 | ||
GenBank mRNA | AF395746.1 | - | 8 | 690 | ||
BC083546.1 | - | 8 | 658 | 7134940 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dclre1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147145.2 | NP_671486.2 | 8 | 658 | ||
GenBank mRNA | AF395746.1 | - | 8 | 690 | ||
BC083546.1 | - | 8 | 658 | 7134940 |