siRNA Details
| Gene Symbol: Rwdd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147146.2 | NP_671487.1 | 7 | 793 | ||
| XM_008772869.1 | XP_008771091.1 | 7 | 925 | |||
| GenBank mRNA | FQ218614.1 | - | 4 | 328 | ||
| FQ220470.1 | - | 4 | 328 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rwdd1| Gene Symbol: Rwdd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147146.2 | NP_671487.1 | 7 | 793 | ||
| XM_008772869.1 | XP_008771091.1 | 7 | 925 | |||
| GenBank mRNA | FQ218614.1 | - | 4 | 328 | ||
| FQ220470.1 | - | 4 | 328 | |||