siRNA Details
| Gene Symbol: Rwdd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147146.2 | NP_671487.1 | 2 | 240 | ||
| XM_008772869.1 | XP_008771091.1 | 2 | 372 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rwdd1| Gene Symbol: Rwdd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147146.2 | NP_671487.1 | 2 | 240 | ||
| XM_008772869.1 | XP_008771091.1 | 2 | 372 | |||