siRNA Details
| Gene Symbol: Osbpl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172023.1 | NP_742020.1 | 21 | 2230 | ||
| GenBank mRNA | AB089316.1 | - | 21 | 2230 | ||
| FQ214328.1 | - | 7 | 853 | |||
| FQ228055.1 | - | 5 | 603 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Osbpl1a| Gene Symbol: Osbpl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172023.1 | NP_742020.1 | 21 | 2230 | ||
| GenBank mRNA | AB089316.1 | - | 21 | 2230 | ||
| FQ214328.1 | - | 7 | 853 | |||
| FQ228055.1 | - | 5 | 603 | |||