siRNA Details
| Gene Symbol: Ppp1r14d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172011.1 | NP_742008.1 | 1 | 97 | ||
| GenBank mRNA | AY122322.1 | - | 1 | 97 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r14d| Gene Symbol: Ppp1r14d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172011.1 | NP_742008.1 | 1 | 97 | ||
| GenBank mRNA | AY122322.1 | - | 1 | 97 | ||