siRNA Details
| Gene Symbol: Ppm1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147209.2 | NP_671742.1 | 6 | 1059 | ||
| GenBank mRNA | AF525687.1 | - | 6 | 935 | ||
| BC062083.1 | - | 6 | 1059 | 5599833 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppm1g| Gene Symbol: Ppm1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147209.2 | NP_671742.1 | 6 | 1059 | ||
| GenBank mRNA | AF525687.1 | - | 6 | 935 | ||
| BC062083.1 | - | 6 | 1059 | 5599833 | ||