siRNA Details
| Gene Symbol: Zfp335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762559.1 | XP_008760781.1 | 18 | 3436 | ||
| XM_008762560.1 | XP_008760782.1 | 19 | 4110 | |||
| XM_008762561.1 | XP_008760783.1 | 18 | 3433 | |||
| XM_008762562.1 | XP_008760784.1 | 18 | 3436 | |||
| XM_008762563.1 | XP_008760785.1 | 18 | 3436 | |||
| XM_008762564.1 | XP_008760786.1 | 18 | 3433 | |||
| XM_008762565.1 | XP_008760787.1 | 18 | 3436 | |||
| GenBank mRNA | AF309071.1 | - | 4 | 458 | ||
| AY079168.1 | - | 10 | 1208 | |||

Zfp335
Genomic Map