siRNA Details
| Gene Symbol: Msi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148890.1 | NP_683688.1 | 3, 4 | 316 | ||
| GenBank mRNA | AY043393.1 | - | 3, 4 | 316 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msi1| Gene Symbol: Msi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148890.1 | NP_683688.1 | 3, 4 | 316 | ||
| GenBank mRNA | AY043393.1 | - | 3, 4 | 316 | ||